Learn More
Medchemexpress LLC N-Propargylglycine | 58160-95-5 | 99.7% | C5H7NO2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY142035
Encompass_Preferred
N-Propargylglycine is a brain-penetrant, orally active PRODH inhibitor. It works by covalently modifying enzyme-bound FAD and active site lysine, leading to enzyme structural distortion, protein decay, and irreversible inhibition of proline and 4-hydroxyproline catabolism. It can induce UPRmt, upregulate mitochondrial chaperones, and enhance mitochondrial proteostasis.
  • Brain-penetrant and orally active.
  • Irreversibly inhibits proline oxidation.
  • Induces mitochondrial unfolded protein response.
  • Blocks astrocytic L-proline consumption.
  • Stimulates neural processes and increases brain amino acid levels.
  • Reduces hyperoxaluria and prevents calcium oxalate stone formation.
  • Used in research for breast cancer, neurodegenerative disorders, and primary hyperoxaluria.
Catalog No. 50-003-54731