Learn More
Cayman Chemical N-PropIonylglycIn 250mg
SDP
Supplier:  Cayman Chemical 34377250 MG
Encompass_Preferred
A glycine-conjugated form of propionic acid; urinary levels are increased in patients with propionic acidemia
Catalog No. 50-294-7042