Learn More
Medchemexpress LLC Nω-Propyl-L-arginine | 137361-05-8 | MFCD01320455 | 98.6% | 216.28 g/mol | C9H20N4O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10206210MG
Encompass_Preferred
Nω-Propyl-L-arginine is a potent, competitive inhibitor of neuronal nitric oxide synthase (nNOS) used as a research tool to study nNOS-mediated nitric oxide signaling. It is a solid powder with reported purity of 98.62%, molecular weight 216.28 g/mol, and the free base chemical formula C9H20N4O2. Store as a powder at -20°C for optimal stability.
  • Selective neuronal nitric oxide synthase (nNOS) inhibitor
  • Reported Ki for nNOS: 57 nM
  • Approximately 149-fold selectivity for nNOS over eNOS
  • High purity suitable for biochemical and cellular studies
  • Solid powder form with recommended cold storage
  • Useful for studies in immunology and inflammation pathways
Catalog No. 50-002-46193