Learn More
Medchemexpress LLC Nω-propyl-L-arginine hydrochloride | 137361-05-8 | MFCD01320455 | 98.6% | 216.28 g/mol | C9H20N4O2 · xHCl | 100 MG
SDP
Supplier:  Medchemexpress LLC HY102062100MG
Encompass_Preferred
Nω-Propyl-L-arginine hydrochloride is a synthetic Nω-substituted L-arginine analogue that functions as a potent, competitive, and selective inhibitor of neuronal nitric oxide synthase (nNOS). It is provided as a research reagent for biochemical and pharmacological studies of nitric oxide signaling and nNOS-related pathways.
  • Potent, competitive nNOS inhibitor.
  • High selectivity for nNOS over eNOS and iNOS.
  • Suitable for biochemical and pharmacological assays.
  • Offered in multiple milligram package sizes for research needs.
  • High purity (typically ≥98.6% by HPLC).
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-45467