Learn More
Medchemexpress LLC N,S-Bis-Fmoc-Glutathione (TFA) | 149438-56-2 | 751.8 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYP4277A50MG
Encompass_Preferred
N,S-Bis-Fmoc-glutathione is a sophisticated glutathione derivative for advanced applications in peptide synthesis and drug development. Its unique structure enhances stability and solubility, suitable for researchers in bioconjugation and targeted therapies. Protective Fmoc groups enable selective deprotection, facilitating precise synthesis. This product is for research use only, intended for laboratory chemicals and substance manufacturing.
  • Acts as a protecting group in peptide synthesis, allowing selective modification of amino acids.
  • Crucial for developing new pharmaceuticals, targeting specific biological pathways.
  • Used to attach biomolecules to surfaces, enhancing drug delivery and diagnostic tools.
  • Enables the study of oxidative stress mechanisms and development of therapeutic strategies.
  • Employed in analytical techniques to improve assay sensitivity and selectivity in biochemical research.
Catalog No. 50-003-97166