Learn More
Medchemexpress LLC N-sec-butyl Pentylone (hydrochloride) | 17763-05-2 | 99.0% | 313.82 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY402939A1MG
Encompass_Preferred
N-sec-butyl Pentylone hydrochloride is a drug derivative intended for research use only and not for sale to patients. It appears as a solid, white to off-white substance.
  • Drug derivative
  • For research use only
  • Not sold to patients
  • Solid, white to off-white appearance
  • Ships at room temperature in continental US (may vary elsewhere)
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture)
Catalog No. 50-004-01433