Learn More
Medchemexpress LLC N-substituted 5-(1H-indol-2-yl)-2-methoxyaniline | 2764818-24-6 | 98.0% | 473.59 | C24H31N3O5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1437875MG
Encompass_Preferred
ALOX15-IN-1 is a small-molecule research inhibitor of ALOX15 (linoleate oxygenase) that potently inhibits rabbit and human enzymes. It is provided as a solid (off-white to light yellow) with high analytical purity, and is characterized by CAS 2764818-24-6, formula C24H31N3O5S, and molecular weight 473.59 g/mol. Recommended storage varies for powder and solutions.
  • Potent inhibitor of ALOX15 with low-nanomolar activity for linoleic acid (IC50 0.04 μM).
  • Detectable activity against arachidonic acid (IC50 2.06 μM).
  • High purity (98.01%) suitable for research applications.
  • Solid, off-white to light yellow, compatible with common analytical methods.
  • Stable under recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months).
  • Available in milligram-scale quantities for laboratory studies.
Catalog No. 50-002-22302