Learn More
Medchemexpress LLC N-succinimidyl 3-(acetylthio)propionate | 84271-78-3 | MFCD00133023 | ≥98.0% | 245.25 g/mol | C9H11NO5S | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0139761G
Encompass_Preferred
3-(Acetylthio)propionic acid N-succinimidyl ester is an N-hydroxysuccinimide (NHS) ester reagent that installs an acetyl-protected thiol onto primary amines. After deacetylation, the resulting thiol is available for conjugation, crosslinking, or linker synthesis in biomolecular and organic chemistry.
  • Introduces an acetyl-protected thiol onto amine-containing molecules for controlled thiol installation.
  • Allows deprotection to reveal free thiols for downstream conjugation and crosslinking.
  • Compatible with peptide and protein modification workflows.
  • Supplied as a stable solid for convenient storage and handling.
  • High purity supports reproducible coupling and reduced side reactions.
Catalog No. 50-003-02289