Learn More
Medchemexpress LLC N-succinimidyl 4-(dimethylamino)benzoate | 58068-85-2 | MFCD16875522 | ≥98.0% | 262.3 g/mol | C13H14N2O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY118431100MG
Encompass_Preferred
DMABA NHS ester (N-succinimidyl 4-(dimethylamino)benzoate, CAS 58068-85-2) is an NHS-ester reagent that reacts with primary amines to form stable amide bonds. It is commonly used to derivatize phosphatidylethanolamine and other amine-containing molecules to improve detection by mass spectrometry and chromatographic methods.
  • Reacts with primary amines to form stable amide bonds.
  • Enhances detectability of phosphatidylethanolamine by mass spectrometry.
  • High purity suitable for analytical and derivatization workflows.
  • Available in small milligram quantities for method development and optimization.
  • Solid form simplifies storage and handling for laboratory use.
Catalog No. 50-002-28551