Learn More
Medchemexpress LLC N-succinimidyl 6-(3-maleimidopropionamido) hexanoate | 367927-39-7 | MFCD01863460 | 95.0% | 379.36 g/mol | C17H21N3O7 | 500MG
SDP
Supplier:  Medchemexpress LLC HY124318500MG
Encompass_Preferred
SMPH crosslinker is a heterobifunctional amine-to-thiol crosslinking reagent containing an N-hydroxysuccinimide (NHS) ester and a maleimide group, used as a linker in targeted protein degradation chemistry and bioconjugation. It is supplied as a white to off-white solid with molecular formula C17H21N3O7 and molecular weight 379.36 g/mol, and is commonly used to connect amine-containing and thiol-containing molecules in synthesis workflows.
  • Contains NHS-ester and maleimide functional groups for orthogonal coupling
  • Enables amine-to-thiol conjugation for bioconjugation and PROTAC assembly
  • White to off-white solid, suitable for standard laboratory storage
  • High purity typically ≥95.0%
  • Supplied in a 500 mg quantity for bench-scale synthesis
Catalog No. 50-002-36868