Learn More
Medchemexpress LLC HY-114176 500mg Medchemexpress, (S,R,S)-AHPC-C4-NH2 (hydrochloride) CAS:2245697-83-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY114176500MG
Encompass_Preferred
Medchemexpress, HY-135233 50mg N-Succinimidyl-S-acetylthioacetate CAS:76931-93-6 N-Succinimidyl S-acetylthioacetate (SATA), a protein modification agent, introduces thiol-groups into protein molecules. N-Succinimidyl S-acetylthioacetate adds sulfhydryl groups to proteins and other amine-containing molecules in a protected form. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9299