Learn More
Medchemexpress LLC N-tert-Boc-cis-4-fluoro-L-proline | 203866-13-1 | 233.24 | 5 G

Supplier: Medchemexpress LLC HYW0032225G
N-tert-Boc-cis-4-fluoro-L-proline is a drug intermediate for the synthesis of 4-fluoropyrrolidine-2-carbonitrile DPP-4 inhibitor. It can be used to study type 2 diabetes.
- Drug intermediate for 4-fluoropyrrolidine-2-carbonitrile DPP-4 inhibitor synthesis
- Suitable for studying type 2 diabetes
- For research use only
- Appearance: white to off-white solid
- Ships at room temperature
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
- Soluble in DMSO (≥ 100 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.