Learn More
Medchemexpress LLC N-(tert-butoxycarbonyl)-beta-alanine | 3303-84-2 | MFCD00037291 | ≥98.0% | 189.21 g·mol⁻¹ | C8H15NO4 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW015231500G
Encompass_Preferred
Boc-β-Ala-OH (N-(tert-butoxycarbonyl)-beta-alanine) is a Boc-protected beta-alanine used as a building block in peptide synthesis and organic chemistry research. It is supplied as a solid reagent with high purity for laboratory use; intended for research use only.
  • Boc-protected beta-alanine suitable for peptide coupling and synthesis workflows.
  • High purity (≥98.0%) for consistent experimental results.
  • Molecular weight 189.21 g·mol⁻¹ and formula C8H15NO4 for analytical reference.
  • Available in larger package sizes to support scale-up of reactions.
  • Certificate of analysis provided to verify batch quality.
Catalog No. 50-003-02558