Learn More
Medchemexpress LLC N-tert-butyl-3-(acetamido)benzamide | 183619-38-7 | 99.8% | 234.29 g·mol⁻¹ | C13H18N2O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1003765MG
Encompass_Preferred
CPI-1189 is a small-molecule research compound described as an orally active inhibitor of tumor necrosis factor-α (TNF-α) release with reported neuroprotective and anti-inflammatory activity. It inhibits p38 MAPK phosphorylation and is used in preclinical studies of apoptosis, ischemia/reperfusion injury, and neurodegenerative disease models.
  • Orally active TNF-α release inhibitor.
  • Inhibits phosphorylation of p38 MAPK.
  • Reported neuroprotective and anti-inflammatory activity.
  • Used in in vitro and in vivo preclinical models of apoptosis and ischemia.
  • High purity suitable for research applications.
  • Stable as powder at -20°C for long-term storage.
Catalog No. 50-002-14218