Learn More
Medchemexpress LLC N-tetradecanoyl-L-homoserine lactone | 202284-87-5 | MFCD28899662 | 99.6% | C18H33NO3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13368410MG
Encompass_Preferred
N-tetradecanoyl-L-homoserine lactone is an N-acyl homoserine lactone quorum-sensing signal used as an analytical standard in bacterial signaling and quorum-sensing research. It is supplied as a highly purified small-molecule standard suitable for biochemical assays, method development, and quantitative analysis.
  • High purity (≈99.6%).
  • Chemical formula C18H33NO3 and molecular weight 311.46 g/mol.
  • Suitable as analytical standard for LC-MS and HPLC method development.
  • Used to study bacterial quorum-sensing and intercellular signaling.
  • Available in small milligram pack sizes for research workflows.
Catalog No. 50-002-60538