Learn More
Medchemexpress LLC N-tridecyl alcohol-d27 | 203633-18-5 | 227.53 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW004293S100MG
Encompass_Preferred
N-Tridecyl Alcohol-d27 is the deuterium labeled 1-Tridecanol. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled 1-Tridecanol
  • Used as a tracer for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Store in pure form at -20°C for 3 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-48847