Learn More
Medchemexpress LLC N-(trifluoroacetyl)aminoacetic acid | 383-70-0 | MFCD00029784 | >98.0% | 171.07 | C4H4F3NO3 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW016948
Encompass_Preferred
TFA-Gly-OH (N-(trifluoroacetyl)glycine) is the trifluoroacetyl derivative of glycine used as a reagent and building block in peptide synthesis and amino-acid derivatization. Supplied as a white to off-white solid for research use, it provides a protected glycine unit for acylation and coupling steps, with recommended storage conditions to preserve long-term stability.
  • Trifluoroacetyl-protected glycine for peptide synthesis and acylation reactions.
  • White to off-white solid, easy to weigh and handle.
  • Stable under recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Suitable for research use and synthetic workflows.
  • Available in multiple pack sizes for scalability.
Catalog No. 50-003-04160