Learn More
Medchemexpress LLC N-(trifluoroacetyl)glycine | 383-70-0 | MFCD00029784 | 171.07 g/mol | C4H4F3NO3 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01694810G
Encompass_Preferred
TFA-Gly-OH (N-(trifluoroacetyl)glycine) is a trifluoroacetyl-protected glycine derivative used as an amino-acid building block and reagent in chemical synthesis and research applications. It is supplied as a white to off-white solid (C4H4F3NO3, MW 171.07 g/mol) and is available in laboratory pack sizes for routine handling.
  • Trifluoroacetyl-protected amino acid suitable for peptide synthesis and derivatization
  • White to off-white solid for straightforward storage and handling
  • Molecular formula C4H4F3NO3 and molecular weight 171.07 g/mol
  • Available in multiple laboratory pack sizes for scalable use
  • Compatible with common synthetic workflows and protecting-group strategies
Catalog No. 50-003-00432