Learn More
Medchemexpress LLC N-valyltryptophan (dipeptide 2) | 24587-37-9 | MFCD00037947 | 97.9% | 303.36 g/mol | C16H21N3O3 | 25 G

Supplier: Medchemexpress LLC HY11806025G
Dipeptide 2 (N-Valyltryptophan) is a bioactive dipeptide used as an anti-aging cosmetic ingredient; it is reported to improve lymphatic circulation and reduce puffiness and eye bags. Supplied as a white powder for research and formulation use.
- CAS 24587-37-9, molecular weight 303.36 g/mol, formula C16H21N3O3.
- Purity approximately 97.9% as provided by manufacturer.
- Soluble in DMSO (~12.5 mg/mL); ultrasonic assistance recommended for dissolution.
- Used in cosmetic formulations to reduce under-eye puffiness and bags.
- Store powder at -20°C (up to 3 years) or 4°C (up to 2 years); store solutions at -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.