Learn More
Medchemexpress LLC N-vanillyloleamide | 58493-49-5 | MFCD00673962 | ≥99.0% | 417.62 g/mol | C26H43NO3 | 5 MG

Supplier: Medchemexpress LLC HY1013235MG
Olvanil (NE-19550) is a small-molecule agonist of transient receptor potential vanilloid 1 (TRPV1) channels with reported analgesic activity. It is used in preclinical research to probe TRPV1-mediated signaling and pain pathways. Key identifiers include chemical formula C26H43NO3, molecular weight 417.62 g/mol, and CAS 58493-49-5.
- Agonist of TRPV1 channels, useful for pain signaling studies.
- High purity ≥99.0%.
- Available as small solid quantities and as a 10 mM DMSO solution.
- Soluble in DMSO at 100 mg/mL with warming or sonication.
- Recommended storage: solid at -20°C under inert gas; in solvent -80°C for long-term storage.
- Includes SDS and COA documentation for handling and quality verification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.