Learn More
Chem-Impex International, Inc. N-Vinyl-2-Pyrrolidone 99%, Versatile Monomer, Liquid Form
Supplier:  Chem-Impex International, Inc. 268634KG
Encompass_Preferred
N-Vinyl-2-pyrrolidone, 99%, 4 kg is a versatile monomer crucial for synthesizing polymers and copolymers. Its excellent solubility in water and organic solvents makes it essential in UV-curable coatings, inks, and adhesives, enhancing their durability and adhesion. Additionally, it acts as a binder in pharmaceuticals, improving active ingredient solubility. The product is stabilized to prevent spontaneous polymerization, ensuring stability during storage and use, setting it apart from other chemicals.
Catalog No. NC3496234