Learn More
Medchemexpress LLC N1-(1,8-dimethylimidazo[1,2-a]quinoxalin-4-yl)ethane-1,2-diamine hydrochloride | 547757-23-3 | MFCD07784499 | >98.0% | 291.78 g/mol | C14H18ClN5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10518100MG
Encompass_Preferred
BMS-345541 hydrochloride is the hydrochloride salt of a selective small-molecule inhibitor targeting the catalytic subunits of IκB kinase (IKK). It binds at an allosteric site and is used as a research reagent to modulate NF-κB signaling in cell and animal studies.
  • Selective inhibition of IKK catalytic subunits (IKK-2 IC50 ≈ 0.3 μM, IKK-1 IC50 ≈ 4 μM).
  • Binds at an allosteric site on IKK.
  • Suitable for in vitro and in vivo studies to modulate NF-κB signaling.
  • White to off-white solid with good solubility in DMSO.
  • Typical purity ≥98.0% (HPLC).
  • Molecular weight 291.78 g/mol.
Catalog No. 50-002-43702