Learn More
TARGETMOL CHEMICALS INC N1-Acetylspermidine hydrochlor
SDP
Supplier:  TARGETMOL CHEMICALS INC T371705MG
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. N1-Acetylspermidine hydrochloride, an acetyl derivative of spermidine, serves as a substrate for polyamine oxidase and selectively elevates its levels in human colorectal adenocarcinomas. Purity 99.76%
Catalog No. 50-287-9344