Learn More
Medchemexpress LLC N1-guanyl-1,7-diaminoheptane sulfate | 150417-90-6 | ≥98.0% | 270.35 | C8H22N4O4S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY108314A10MM 1ML
Encompass_Preferred
GC7 sulfate is a deoxyhypusine synthase (DHPS) inhibitor supplied for research use. It is offered as ready-to-use 10 mM solutions and as solids for preparation of stock solutions, and is commonly used in biochemical and cellular studies targeting hypusination-dependent pathways.
  • Potent inhibitor of deoxyhypusine synthase (DHPS)
  • Available as 10 mM solution and multiple solid sizes for flexible use
  • Molecular formula C8H22N4O4S; molecular weight approximately 270.35 g/mol
  • Suitable for biochemical and cellular research applications
  • Datasheet provides guidance for stock solution preparation
Catalog No. 50-002-42496