Learn More
Medchemexpress LLC N1-Methylinosine | 2140-73-0 | 99.7% | 282.25 | 1 ML

Supplier: Medchemexpress LLC HY11313910MM/1ML
1-Methylinosine is a modified nucleotide found at position 37 in tRNA 3' to the anticodon of eukaryotic tRNA. It is a metabolite that can be derived from inosine, and its levels are notably higher in urine samples from breast cancer models. It is available as a solution in DMSO.
- Target: Nucleoside antimetabolite/analog; endogenous metabolite
- Pathway: Cell cycle/DNA damage; metabolic enzyme/protease
- Appearance: White to off-white solid
- Solubility in DMSO: 83.33 mg/mL (295.23 mM)
- Storage (powder): -20°C for 3 years, 4°C for 2 years
- Storage (in solvent): -80°C for 2 years, -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.