Learn More
Medchemexpress LLC N1,N1-Dimethylpropane-1,2-diamine dihydrochloride | 75975-34-7 | MFCD16619104 | ≥96.0% | 175.10 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY32007A
Encompass_Preferred
N1,N1-Dimethylpropane-1,2-diamine dihydrochloride is a chemical compound primarily used as a drug intermediate for the synthesis of various active compounds. It is intended for research use only and is not sold to patients. This product has specific storage requirements to maintain its stability and efficacy.
  • Used as a chemical intermediate in drug synthesis.
  • Intended for research applications.
  • Requires storage at -20°C for up to 3 years as a powder.
  • In solvent, it requires storage at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-004-03244