Learn More
Cayman Chemical N1 N12-DIacetylspermInhydr 5mg
SDP
Supplier:  Cayman Chemical 179185 MG
Encompass_Preferred
A diacetylated derivative of spermine that displays lower Km and higher Vmax values than spermine, making it a better substrate of polyamine oxidase than the non-acetylated polyamine; used as a biomarker for cancer detection
Catalog No. 50-296-7487