Learn More
Medchemexpress LLC N1,n3-bis((r)-4-((4-((3,5-dibromobenzyl)oxy)phenyl) sulfonamido)-5-(hydroxyamino)-5-oxopentyl)isopht | 2758892-64-5 | 98.2% | C44H44Br4N6O12S2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13291710MG
Encompass_Preferred
This small-molecule ADAM8 inhibitor is designed for research applications to probe ADAM8-mediated pathways. It shows potent biochemical activity and is supplied with quality documentation for experimental use.
  • Potent ADAM8 inhibitor with an IC50 of 73 nM.
  • High purity: 98.2% (HPLC, batch COA).
  • Molecular formula C44H44Br4N6O12S2; molecular weight 1232.60 g/mol.
  • Provided in small pack sizes suitable for screening and validation studies.
  • Supplied with certificate of analysis and safety data sheet documentation.
  • Suitable for biochemical and cell-based assays for target validation.
  • Intended for research use only.
Catalog No. 50-002-61496