Learn More
Medchemexpress LLC N2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N6-trityl-L-lysine | 111061-54-2 | MFCD00270545 | 98.6% | 610.74 g·mol⁻¹ | C40H38N2O4 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW048677
Encompass_Preferred
N2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N6-trityl-L-lysine is an Fmoc- and trityl-protected L-lysine derivative used as a protected amino acid building block for solid-phase and solution-phase peptide synthesis and related synthetic chemistry research.
  • Fmoc- and trityl-protected lysine for orthogonal side-chain protection.
  • High purity suitable for peptide coupling and synthesis.
  • Available in common laboratory pack sizes, including 25 G.
  • Certificate of analysis (COA) and safety data sheet (SDS) available.
  • Molecular weight 610.74 g·mol⁻¹; formula C40H38N2O4.
Catalog No. 50-002-09499