Learn More
Medchemexpress LLC N2-Fmoc-N6-(5-dimethylaminonaphthalen-1-ylsulfonyl)-L-lysine | 118584-90-0 | MFCD00672343 | 96.4% | 601.71 g·mol⁻¹ | C33H35N3O6S | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0537011G
Encompass_Preferred
N2-(((9H-fluoren-9-yl)methoxy)carbonyl)-N6-((5-(dimethylamino)naphthalen-1-yl)sulfonyl)-L-lysine is an Fmoc-protected lysine derivative bearing a dansyl-type fluorescent sulfonyl group, supplied as a light yellow solid for research use.
  • Fmoc-protected on the α-amino (N2) for standard peptide synthesis workflows.
  • N6-substituted with a 5-(dimethylamino)naphthalen-1-yl sulfonyl group for fluorescence labeling.
  • Suitable as a fluorescent amino acid building block in peptide labeling and analytical studies.
  • High purity (96.38%) and light yellow appearance facilitate analytical characterization.
  • Stable when stored as powder at -20°C for long-term storage.
Catalog No. 50-002-10030