Learn More
TARGETMOL CHEMICALS INC N2-Methylguanosine 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T75145MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. N2-Methylguanosine is a modified nucleoside found at several specific sites in various tRNAs. Purity 98%
Catalog No. 50-287-8018