Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N2,N6-bis(((9H-fluoren-9-yl)methoxy)carbonyl)-D-lysine | 75932-02-4 | 95.7% | 590.66 g/mol | C36H34N2O6 | 100 G
SDP

Supplier:  Medchemexpress LLC HYW048691100G

Encompass_Preferred

N2,N6-Bis(((9H-fluoren-9-yl)methoxy)carbonyl)-D-lysine is an Fmoc-protected D-lysine derivative used as a building block in peptide synthesis and other organic synthesis applications.

  • Fmoc-protected D-lysine derivative suitable for peptide synthesis.
  • Purity 95.7% as supplied.
  • Molecular formula C36H34N2O6; molecular weight 590.66 g/mol.
  • Appearance: white to off-white solid.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Available in gram-scale quantities including 100 G.

Catalog No. 50-002-09876


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.