Learn More
Medchemexpress LLC N3-C2-NHS ester | 850180-76-6 | MFCD24452814 | 99.8% | 212.16 g/mol | C7H8N4O4 | 25 MG

Supplier: Medchemexpress LLC HY12652025MG
N3-C2-NHS ester is an azide-containing, noncleavable NHS-activated linker used for bioconjugation in antibody-drug conjugate synthesis and other click-chemistry applications. It couples to primary amines via NHS-ester chemistry and participates in CuAAC and strain-promoted azide-alkyne cycloaddition reactions.
- Azide-containing linker suitable for CuAAC and SPAAC reactions.
- NHS ester allows covalent coupling to primary amines.
- High purity suitable for research use.
- Soluble in DMSO for stock solution preparation (100 mg/mL).
- Recommended storage: solid at -20 °C; in solution at -80 °C or -20 °C depending on duration.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.