Learn More
Medchemexpress LLC N3-C3-NHS ester | 943858-70-6 | MFCD22573606 | 99.9% | 226.19 g/mol | C8H10N4O4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY126521500MG
Encompass_Preferred
N3-C3-NHS ester is an azide-containing N-hydroxysuccinimide (NHS) ester used as a noncleavable linker and click-chemistry reagent for bioconjugation and antibody-drug conjugate (ADC) synthesis. It installs an azide handle that reacts in copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC) with alkyne, DBCO, or BCN partners.
  • Noncleavable ADC linker used in antibody-drug conjugate synthesis.
  • Azide functional group enables CuAAC and SPAAC click chemistries.
  • Reactive NHS ester couples to primary amines on biomolecules.
  • Powder form; soluble in DMSO for conjugation workflows.
  • High purity (99.85%) and defined molecular weight (226.19 g/mol).
Catalog No. 50-002-03456