Learn More
Medchemexpress LLC N3-Gly-Gly-Gly-Gly-Gly-OH | 2250433-77-1 | C10H15N7O6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1517775MG
Encompass_Preferred
N3-Gly-Gly-Gly-Gly-Gly-OH is a click chemistry reagent that contains an azide group. It is capable of participating in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing alkyne groups. Additionally, it can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Undergoes ring strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-73284