Learn More
Medchemexpress LLC N3-Gly-Gly-OH | 855750-87-7 | C4H6N4O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY151859
Encompass_Preferred
N3-Gly-Gly-OH is a click chemistry reagent utilized in various research fields due to its high yield, high specificity, and simplicity in binding nucleic acids, lipids, proteins, and other molecules. This reagent contains an azide group, enabling its participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, as well as ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Click chemistry reagent
  • High yield
  • High specificity
  • Simplicity in binding
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in ring strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Useful for binding nucleic acids, lipids, proteins, and other molecules
  • Applicable in many research fields
Catalog No. 50-003-74233