Learn More
Medchemexpress LLC N3-L-Val-OH (CHA) | 1217462-63-9 | C11H22N4O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY151661
Encompass_Preferred
N3-L-Val-OH (CHA) is a click chemistry reagent containing an azide group and a valine derivative. It can serve as a synthetic intermediate for Valaciclovir (Valacyclovir). It participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Contains an azide group and is a valine derivative
  • Can serve as a synthetic intermediate for valaciclovir (valacyclovir)
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Participates in ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups
Catalog No. 50-003-74414