Learn More
Medchemexpress LLC N3-PEG11-CH2CH2Br | 2098982-00-2 | MFCD32696701 | >98.0% | 634.56 g/mol | C24H48BrN3O11 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW190881100MG
Encompass_Preferred
N3-PEG11-CH2CH2Br is an azide-terminated polyethylene glycol (PEG11) linker bearing a terminal bromoethyl group. Supplied as a high-purity research reagent (purity >98%), it is intended for bioconjugation, probe synthesis, and other linker chemistry where azide reactivity and alkyl bromide substitution are required. The hydrophilic PEG spacer enhances solubility in aqueous media, facilitating handling in biochemical assays. Typical pack size: 100 mg.
  • Azide-terminated linker for click chemistry.
  • Terminal bromoethyl group enables nucleophilic substitution.
  • PEG11 spacer improves aqueous solubility.
  • High purity suitable for research applications.
  • Supplied in small-scale vials for synthesis and probe development.
Catalog No. 50-002-81939