Learn More
Medchemexpress LLC N3-PEG3-C2-NHS ester | 1245718-89-1 | 99.9% | 344.32 g/mol | C13H20N4O7 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1265281G
Encompass_Preferred
N3-PEG3-C2-NHS ester is an azide-terminated PEG3 linker bearing an N-hydroxysuccinimide (NHS) ester reactive group, designed for coupling to primary amines and for use in click-chemistry workflows such as antibody-drug conjugate (ADC) synthesis.
  • Azide-terminated PEG3 linker for click chemistry applications.
  • NHS ester reactive handle for coupling to primary amines.
  • Non-cleavable linker suitable for stable bioconjugation.
  • Reported high chemical purity for research use.
  • Available in multiple pack sizes including 1 G for laboratory-scale synthesis.
Catalog No. 50-002-02422