Learn More
Medchemexpress LLC N3-PEG3-C2-NHS ester | 1245718-89-1 | 99.9% | 344.32 g/mol | C13H20N4O7 | 500 MG

Supplier: Medchemexpress LLC HY126528500MG
N3-PEG3-C2-NHS ester is a non-cleavable three-unit polyethylene glycol (PEG3) linker bearing an azide functional group and an N-hydroxysuccinimide (NHS) ester. It is used as a click-chemistry reagent and linker in the synthesis of antibody-drug conjugates and other bioconjugation applications.
- Non-cleavable three-unit PEG linker for stable conjugation.
- Contains azide and NHS ester functional groups for click chemistry and amine coupling.
- High purity suitable for research applications.
- Soluble in DMSO at high concentration; requires ultrasonic treatment.
- Supplied as a colorless to light yellow liquid; multiple pack sizes available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.