Learn More
Medchemexpress LLC N3-PEG3-CH2CH2COOH | 1056024-94-2 | ≥98.0% | 247.25 g/mol | C9H17N3O5 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY42490250MG
Encompass_Preferred
N3-PEG3-CH2CH2COOH is an azido-terminated, triethylene glycol (PEG3) propionic acid linker reagent used in chemical synthesis for conjugation and click chemistry applications. It provides an azide functional group suitable for copper-catalyzed and strain-promoted azide-alkyne cycloadditions, and a carboxylic acid handle for covalent coupling.
  • Azide functional group for CuAAC and SPAAC reactions.
  • Triethylene glycol spacer for increased solubility and flexibility.
  • Carboxylic acid handle for straightforward conjugation chemistry.
  • High purity for research use.
  • Stable under recommended storage: -20°C, protect from light; in solvent -80°C for up to 6 months.
Catalog No. 50-002-02763