Learn More
Medchemexpress LLC N3-peg3-vc-pab-mmae | 99.1% | 1352.66 | C67H109N13O16 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10087450MG
Encompass_Preferred
N3-PEG3-vc-PAB-MMAE is a synthesized drug-linker conjugate combining an azide-functionalized PEG3 spacer, a valine-citrulline-para-aminobenzyl (vc-PAB) cleavable linker, and the cytotoxic payload monomethyl auristatin E (MMAE). It is provided as a high-purity research reagent for antibody-drug conjugate development and click-chemistry functionalization.
  • Contains azide handle for click chemistry.
  • Includes PEG3 spacer for improved solubility.
  • Features vc-PAB cleavable linker for intracellular release.
  • Carries MMAE tubulin inhibitor payload.
  • Provided with high reported purity.
  • Available in small-scale sizes for research use.
Catalog No. 50-002-11731