Learn More
Medchemexpress LLC N3-PEG3-VC-PAB-MMAF | 2757277-32-8 | 100.0% | 1352.62 g/mol | C66H105N13O17 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15373910MG
Encompass_Preferred
N3-PEG3-VC-PAB-MMAF is an azide-functional drug-linker conjugate used for antibody-drug conjugate (ADC) research and bioconjugation. It combines a PEG3-VC-PAB cleavable linker with the tubulin inhibitor MMAF, and is designed for click-chemistry attachment to alkyne or strained-alkyne partners.
  • Enables click chemistry conjugation via azide handle.
  • Contains a cleavable valine-citrulline-PAB linker for payload release.
  • Suitable for ADC linker-payload synthesis and site-specific attachment.
  • Solid, white to off-white material with high purity for research use.
Catalog No. 50-002-04454