Learn More
Medchemexpress LLC N3-PEG4-C2-NHS ester | 944251-24-5 | 98.9% | 1 G
SDP
Supplier:  Medchemexpress LLC HY1301091G
Encompass_Preferred
N3-PEG4-C2-NHS ester is a non-cleavable 4-unit PEG linker used in the synthesis of antibody-drug conjugates (ADCs). It is a click chemistry reagent containing an azide group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Non-cleavable 4-unit PEG linker
  • Used in the synthesis of antibody-drug conjugates (ADCs)
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-40065