Learn More
Medchemexpress LLC N3-PEG4-C2-NHS ester | 944251-24-5 | 98.9% | 5 G
SDP
Supplier:  Medchemexpress LLC HY1301095G
Encompass_Preferred
N3-PEG4-C2-NHS ester is a non-cleavable 4-unit PEG linker utilized in the synthesis of antibody-drug conjugates (ADCs). It functions as a click chemistry reagent, containing an Azide group. This allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules that contain Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Non-cleavable 4-unit PEG linker
  • Used in synthesis of antibody-drug conjugates (ADCs)
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-40057