Learn More
Medchemexpress LLC N3-PEG4-C2-NHS ester | 944251-24-5 | 98.9% | 500 MG
SDP
Supplier:  Medchemexpress LLC HY130109500MG
Encompass_Preferred
N3-PEG4-C2-NHS ester is a non-cleavable 4-unit PEG linker designed for the synthesis of antibody-drug conjugates (ADCs). This versatile click chemistry reagent features an Azide group, which allows it to readily engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. Furthermore, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions when combined with molecules containing DBCO or BCN groups, offering flexibility in conjugation strategies. This product is suitable for research applications.
  • Non-cleavable PEG linker for stable conjugation.
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions.
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Designed for the synthesis of antibody-drug conjugates (ADCs).
  • Intended for research use only.
Catalog No. 50-003-39037