Learn More
Medchemexpress LLC N3-PEG4-C2-NHS ester | 944251-24-5 | MFCD13184948 | 98.9% | 388.37 | C15H24N4O8 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY13010925MG
Encompass_Preferred
N3-PEG4-C2-NHS ester is a non-cleavable 4-unit polyethylene glycol (PEG) linker containing an azide group and an N-hydroxysuccinimide (NHS) ester for amine coupling; it is used for bioconjugation and click chemistry applications such as antibody-drug conjugate synthesis.
  • Azide functional group enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC).
  • NHS ester reacts with primary amines to form stable amide bonds for labeling.
  • Four-unit PEG spacer increases solubility and reduces steric hindrance.
  • High purity suitable for research-grade bioconjugation workflows.
  • Stable when stored under recommended low-temperature conditions to preserve reactivity.
Catalog No. 50-002-02613