Learn More
Medchemexpress LLC N3-TEMPO | 63697-61-0 | C9H17N4O* | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1518475MG
Encompass_Preferred
N3-TEMPO is a click chemistry reagent that contains an azide group. It is useful for binding between nucleic acids, lipids, proteins, and other molecules due to its high yield, specificity, and simplicity. It can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Click chemistry reagent
  • Azide compound
  • Suitable for antibody-drug conjugate/ADC related research
  • Can be used as an ADC linker
Catalog No. 50-003-73259