Learn More
Cayman Chemical N3hydroxydecnoylLHomoser 10mg
SDP
Supplier:  Cayman Chemical 900114710 MG
Encompass_Preferred
A bacterial quorum-sensing signaling molecule; induces expression of tda, a gene with roles in the production of the antibiotic TDA and the algicide roseobacticide, in P. inhibens
Catalog No. 50-289-5972