Learn More
Cayman Chemical N3oxodecnoylLHomoserIn 10mg
SDP
Supplier:  Cayman Chemical 1302610 MG
Encompass_Preferred
A quorum-sensing signaling molecule that is produced by bacteria in response to increasing cell density; increases tra expression in A. tumefaciens in a reporter cell assay; induces adventitious root formation and increases expression of AUX22c, AUX22d, CDC2, ARC, and CDPK in mung bean seedlings at 100 nM; decreases expression of glucanase genes in A. thaliana at 6 µM
Catalog No. 50-295-3261